Specifications
| For Use With (Application) | Mcolortest™ Total Hardness Test in Freshwater |
| No. of Tests | 50 |
| Range | 1 drop corresponds to 1.25°e |
| Test Method | Titrimetric w/Dropping Bottle |
| Type | Total Hardness Test Kit |