Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human MCP-4/CCL13 ELISA Kit

Catalog No. EHCCL13
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHCCL13 96 Tests
EHCCL13X5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHCCL13 Supplier Invitrogen™ Supplier No. EHCCL13
Add to Cart
Add to Cart

ELISA

The Human Monocyte Chemoattractant Protein-4/CCL1 (Hu MCP-4) ELISA quantitates Hu MCP-4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MCP-4. Principle of the method The Human MCP-4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99616
Assay Range 3.13 to 200 pg/mL
Assay Sensitivity 3 pg/mL
Calibration Range 3 pg/mL to 200 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Chemokine & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, ENA-78, FGF- 4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Sufficient For 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 6357
Gene Symbol CKb10,MCP-4,NCC-1,NCC1,SCYA13,SCYL1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym CKb10, MCP-4, NCC-1, NCC1, SCYA13, SCYL1
Target MCP-4
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.