Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCPT-1 (mMCP-1) Monoclonal Antibody (RF6.1), eBioscience™, Invitrogen™

Catalog No. 50245559
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-559 25 μg
50-245-560 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-245-559 Supplier Invitrogen™ Supplier No. 14550380
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The monoclonal antibody RF6 recognized MCPT-1 (Mast Cell Protease-1) which is a β-chymase, a type of serine protease stored and secreted in a tissue-specific manner by mucosal mast cells. MCPT-1 is a chymotryptic protease, referring to its ability to cleave proteins and peptides after an aromatic amino acid. It shares 74% sequence homology with its rat counterpart, rat Mast Cell Protease-II (rMCP-II) and has no direct human counterpart. MCPT-1 is the only chymase expressed by intestinal mucosal mast cells, which are found in the intestinal epithelium. Although it is expressed constitutively and is detectable in the sera of normal mice, parasites in the gut cause systemic levels to increase dramatically within two days and peak at two weeks following infection. It plays an important role in host defense against intestinal parasites, as mice deficient in MCPT-1 display delayed response upon infection. Elevated MCPT-1 levels are also observed during intestinal allergic hypersensitivity reactions. The mechanism of its action is not fully understood, although it is believed to increase intestinal permeability similarly to rMCP-II, possibly by cleaving the proteins at tight junctions between cells. Applications Reported: This RF6 antibody has been reported for use in immunocytochemistry and ELISA. Applications Tested: This RF6 antibody has been tested by ELISA.

MCPT-1 is the only chymase expressed by intestinal mucosal mast cells, which are found in the intestinal epithelium. Although it is expressed constitutively and is detectable in the sera of normal mice, parasites in the gut cause systemic levels to increase dramatically within two days and peak at two weeks following infection. It plays an important role in host defense against intestinal parasites, as mice deficient in MCPT-1 display delayed response upon infection. Elevated MCPT-1 levels are also observed during intestinal allergic hypersensitivity reactions. The mechanism of its action is not fully understood, although it is believed to increase intestinal permeability similarly to rMCPT-II, possibly by cleaving the proteins at tight junctions between cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCPT-1 (mMCP-1)
Applications ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone RF6.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Mcpt1
Gene Accession No. P11034
Gene Alias AV080368; mast cell protease 1; Mcp-1; Mcpt1; mMCP-1
Gene Symbols Mcpt1
Host Species Rat
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17224
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.