Description
- Strong yet lightweight design
- Single lens design with integral side shields for unobstructed view
- Non-Slip soft temple material
- Exclusive Duramass scratch resistant coating
- Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
- ANSI Z87+ and CSA Z94.3
Specifications
Specifications
| Certifications/Compliance | Meets ANSI Z87.1 |
| Lens Tint | Gray |
| Frame Color | Frost Black |
| No. per Case | 144/Cs. |
| Temple Style | Flexible; non-slip rubber head grips |
| Temple Color | Black |
| Lens Material | Polycarbonate |