Learn More
MCR Safety Checkmate™ Safety Glasses
Super-lightweight Checkmate™ provides near total eye orbital seal and features flexible bayonet temples and universal nosebridge, ensuring compliance and comfort for a variety of wearers. Temple is notched at the end to accommodate a break away cord.
$2.95 - $55.70
Specifications
Temple Style Bayonet
View More Specs
Products 5
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
Catalog Number Mfr. No. Lens Tint Lens Coating Price Quantity & Availability  
19-500-207
View Documents
MCR Safety
CK112
Grey
Each for $2.95
Pack of 12 EA for $32.75
Only null left
Add to Cart
19-500-208
View Documents
MCR Safety
CK117
Silver Mirror
Each for $4.40
Pack of 12 EA for $50.00
Only null left
Add to Cart
19-500-210
View Documents
MCR Safety
CK119
Clear Mirror, Indoor/Outdoor
Pack of 12 for $55.70
Only null left
Add to Cart
19-500-205
View Documents
MCR Safety
CK100
Clear Uncoated N/A N/A N/A
19-500-206
View Documents
MCR Safety
CK110
Clear N/A N/A N/A
Description
Description
  • Polycarbonate lens provides near total orbital seal
  • Flexible bayonet temples
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Specifications
Specifications
Bayonet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.