Learn More
MCR Safety Classic PVC/Polyester Jackets
Catalog No. 19060477 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
3X-Large
4X-Large
5X-Large
6X-Large
Hood Type:
Attached
Detachable
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hood Type
19-060477 3X-Large Attached
19-818-224 5X-Large Detachable
19-818-171 4X-Large Detachable
19-810-417 6X-Large Detachable
19-832-004 Small Detachable
19-832-005 Medium Detachable
19-832-006 3X-Large Detachable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-060477 Supplier MCR Safety Supplier No. 200JHX3
Add to Cart
Add to Cart
This item is not returnable. View return policy
Classic Series Material combines the strength and waterproof qualities of PVC with the durability of a polyester backing, making it the most popular in the industry.
  • .35 mm PVC on polyester
  • Attached hood with drawstring Rounded collar of same material 2 patch pockets on the front Snap storm fly front
Specifications
Size 3X-Large
Material Polyester, PVC
Color Yellow
Hood Type Attached
No. of Pockets 2
Size (English) Chest 63 in.
Size (Metric) Chest 3.9 cm