Learn More
MCR Safety Crews™ Storm™ Safety Glasses
Catalog No. 19003045 Shop All MCR Safety Products
Change view
Click to view available options
Frame Color:
Black
Navy
Red
Lens Tint:
Clear
Gray
Lens Coating:
Scratch-resistant
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint Lens Coating
19-003-045 Black Gray -
19-003-044 Black Clear Scratch-resistant
19-003-047 Navy Clear Scratch-resistant
19-003-050 Red Clear Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-003-045 Supplier MCR Safety Supplier No. ST112
Add to Cart
Add to Cart
This item is not returnable. View return policy
Workers in the Storm™ can expect extreme comfort with this face-flattering protective eyewear from Crews™. Ratchet action temples combined with non-slip rubber head grips create a customized fit for virtually any head size.
  • Available in a variety of frame colors and lens combinations
  • Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection
  • Single wraparound dual focal point lens with integral side shields
  • 5 position ratchet action bayonet temples
  • Exclusive Duramass™ scratch resistant coating
  • Non-slip, soft temple material
Compliance
Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Specifications
Certifications/Compliance Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Lens Tint Gray
Frame Color Black
Temple Style Ratchet action
Lens Material Polycarbonate