Learn More
MCR Safety Crews™ Tacoma™ Safety Glasses
Catalog No. 19033413G Shop All MCR Safety Products
Change view
Click to view available options
Frame Color:
Black
Blue
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Gray
Size:
Standard
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint Size
19-033-413G Black Scratch-resistant Gray Standard
19-040-866 Black Scratch-resistant Clear X-Large
19-033-413C Black Scratch-resistant Clear Standard
19-033-412C Blue Scratch-resistant Clear Standard
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-033-413G Supplier MCR Safety Supplier No. TC112
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
With the MCR Safety Tacoma™, you get low cost protection without giving up quality, safety, comfort or durability. It's the same quality you expect from Crews, priced to fit even the tightest budget constraints.
Made of strong, lightweight, scratch-resistant polycarbonate, the one piece wrap-around lens delivers a wide field of vision and lateral impact protection. The molded-in nose bridge comfortably fits a wide variety of faces.
  • Spatula temple with extendable adjustment
  • Single lens design
  • XL version features a lens that fits over most prescription glasses and a 5 position ratchet
  • Meets the ANSI Z87.1 Standard
Order Info
The Tacoma is also available with uncoated lenses and nonadjustable temples for even greater savings.
Specifications
Certifications/Compliance Meets ANSI Z87.1 Standard
Lens Coating Scratch-resistant
Description Scratch-Resistant Coating
Lens Tint Gray
Frame Color Black
Size Standard
Style Wrap-Around Lens Protection
Weight (Metric) 50 g
Lens Material Polycarbonate
UV Protection UV Protection