Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Cut Pro™ 92743PU HyperMax™ Gloves, Polyurethane Coating

Catalog No. 19812631 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-812-631 Large
19-812-629 Small
19-812-630 Medium
19-812-632 X-Large
19-812-633 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-812-631 Supplier MCR Safety Supplier No. 92743PUL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Seamless salt and pepper colored HPPE/synthetic shell that offers cut, tear and abrasion resistance. Gray polyurethane coated palm and fingertips provide a durable grip and hides dirt, grit and grime.

  • 13-gauge seamless salt & pepper colored HPPE/synthetic shell
  • Gray PU palm and fingers
  • Comfortable and easy to wear
  • Long-lasting dexterity

Specifications

ANSI Abrasion Rating ANSI 5
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A5
Material HPPE
Product Type Cut-resistant Gloves
Size Large
For Use With (Application) Automotive Assembly, Material Handling, Landscaping, Glass Handling
Glove Cuff Style Knit
Hem Color Multi-colored
Model 92743PUL
Series Cut Pro™ 92743PU
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.