Learn More
MCR Safety Desperado™ Crews™ Safety Glasses
Catalog No. 19810122 Shop All MCR Safety Products
Change view
Click to view available options
Lens Tint:
Clear Mirror, Indoor/Outdoor
Catalog No. Lens Tint
19-810-122 Clear Mirror, Indoor/Outdoor
1 options
This item is not returnable. View return policy
Catalog No. 19-810-122 Supplier MCR Safety Supplier No. DS119
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Get the look and the protection you deserve with Desperado™ safety glasses from MCR Safety. The Desperado's single lens design provides two distinct optical centers and provides powerful protection even in the most challenging circumstances.
  • Single wrap around dual focus lens
  • Non-slip, soft temple material
  • Bayonet temples
  • Soft, secure TPR nose pad
  • Exclusive Duramass scratch resistant coating
Specifications
Certifications/Compliance Meets ANSI Z87.1 standard
Lens Tint Clear Mirror, Indoor/Outdoor
Frame Color Black
Temple Style Bayonet