Learn More
MCR Safety Indirect Vent Goggles
The body design of the Verdict™ provides a wide, unobstructed field of vision and can be worn with most half-mask respirators. Transparent protective PVC body permits a comfortable fit over prescription eyewear, and the optional soft foam lining cushions the face and improves the seal.
Supplier:  MCR Safety 2400F
 View more versions of this product
Catalog No. 18-999-4572
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Offered in both scratch-resistant polycarbonate lens and anti-fog lens, the Verdict is also available with either a wide elastic headband or a neoprene headband.
  • Impact Resistant
  • Fits over Prescription Glasses
  • Optional Foam Lining
  • Indirect Ventilation
  • ANSI Z87+ and CSA Z94.3
Compliance
Meets ANSI Z87.1-2015, ANSI D3, ANSI D4 and CSA Z94.3 standards.
Specifications
Specifications
Scratch-resistant
Clear
Smoke
Indirect
PVC
ANSI Z87.1
Safety Goggles
Adjustable headband
Polycarbonate
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.