Learn More
MCR Safety Cut Pro™ 9672 Dyneema™ Cut-Resistant Gloves, Polyurethane Coating
Catalog No. 19813603 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-603 Small
19-813-604 Medium
19-813-605 Large
19-813-606 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-813-603 Supplier MCR Safety Supplier No. 9672S
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cut-resistant gloves constructed with Dyneema™ micro-particle boosted high-strength fiber offers cool-touch comfort and maximum durability.
  • 13-gauge salt and pepper DSM Certified Dyneema™ shell
  • Cool and comfortable
  • Cut-resistant, abrasion resistant, exceptional dexterity
  • Gray polyurethane coated palm and fingertips
  • Treated with ActiFresh™
Specifications
ANSI Abrasion Rating ANSI 5
Coating Material Polyurethane
Color Gray, White
Cut Resistance Rating ANSI A2
Material Dyneema
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Abrasive applications, assembly, brick handling, construction, farming, general maintenance, landscaping, plumbing, recycling, shipping/receiving
Glove Cuff Style Knit
Hem Color White
Model 9672S
Series Cut Pro™ 9672
Yarn Gauge 13