Learn More
MCR Safety Cut Pro™ 9672 Dyneema™ Cut-Resistant Gloves, Polyurethane Coating
Catalog No. 19813604 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-604 Medium
19-813-603 Small
19-813-605 Large
19-813-606 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-813-604 Supplier MCR Safety Supplier No. 9672M
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cut-resistant gloves constructed with Dyneema™ micro-particle boosted high-strength fiber offers cool-touch comfort and maximum durability.
  • 13-gauge salt and pepper DSM Certified Dyneema™ shell
  • Cool and comfortable
  • Cut-resistant, abrasion resistant, exceptional dexterity
  • Gray polyurethane coated palm and fingertips
  • Treated with ActiFresh™
Specifications
ANSI Abrasion Rating ANSI 5
Color Gray, White
Cut Resistance Rating ANSI A2
Material Dyneema, Polyethylene
Product Type Cut-resistant Gloves
Size Medium
For Use With (Application) Abrasive applications, assembly, box handling, brick handling, construction, cut protection, farming, general maintenance, knife handling, landscaping, plumbing, recycling
Glove Cuff Style Knit
Hem Color White
Model 9672M
Series Cut Pro™ 9672
Yarn Gauge 13