Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Ninja™ Flex Latex Coated Nylon Work Gloves

Catalog No. 19813622 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-813-622 X-Large
19-813-621 Small
19-813-620 Medium
19-813-650 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-813-622 Supplier MCR Safety Supplier No. N9680XL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Latex coating on the palm and fingers enhances grip capabilities, while Actifresh™ treatment prevents odors and bacteria.

  • 15 gauge nylon shell is comfortable and flexible
  • Gray latex coating on the palm and fingertips enhances grip capabilities
  • Component materials comply with all relevant federal regulations for food contact

Specifications

ANSI Abrasion Rating ANSI 3
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Latex
Color Red, Gray
Exterior Finish Fully Textured
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size X-Large
For Use With (Application) Automotive Assembly, Agriculture, Material Handling, and Construction
Glove Cuff Style Knit
Series Ninja™ N9680
Yarn Gauge 15
Length (English) 10.4 in.
Length (Metric) 26.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.