Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Predalite™ Premium Grade Light Nitrile Coated Work Gloves

Catalog No. 19050604 Shop All MCR Safety Products
Change view
Click to view available options
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-050-604 Medium
19-050-605 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-050-604 Supplier MCR Safety Supplier No. 9780M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves feature a comfortable interlock cotton liner treated with ActiFresh™ to reduce odors and eliminate bacteria. Nitrile coating on the palm and fingers enhances grip capabilities and abrasion resistance.

  • Light nitrile coated palm
  • Soft interlock cotton lining
  • Treated with ActiFresh™ to reduce odors
  • Knit wrist helps prevent dirt and debris from entering the glove
  • All component materials comply with all federal regulations for food contact

Specifications

Coating Material Nitrile
Color Blue
Exterior Finish No Texture
Grade Premium
Material Cotton
Product Type Dip-coated Gloves
Size Medium
For Use With (Application) Assembly, Carpentry, General Manufacturing, and Gardening
Glove Cuff Style Knit
Series 9780
Length (English) 10.5 in.
Length (Metric) 26.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.