Learn More
MCR Safety Regular Weight Natural Cotton/Polyester Gloves with PVC Dots (2 Sides)
Catalog No. 19810333 Shop All MCR Safety Products
Change view
Click to view available options
Size:
X-Small
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-333 X-Small
19-048-595B Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-810-333 Supplier MCR Safety Supplier No. 9660XSM
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
This item is not returnable. View return policy
PVC dots on both sides of the glove for increased grip, and abrasion resistance. Ideal for automotive, assembly line, construction, and farming applications.
  • 7 Gauge Cotton/Polyester Blend
  • PVC Dots provide abrasion resistance and grip
Specifications
Construction String Knit
Coating Material PVC Dots (2 Sides)
Color Natural
Hand Ambidextrous
Material Polycotton
Product Type String Knit Gloves
Size X-Small
Style Reversible
Glove Cuff Color Natural
Glove Cuff Style Knit
Packaging Type Polybag
Yarn Gauge 7
Length (English) 8.5 in.
Length (English) Cuff 2 in.
Length (Metric) 21.6 cm
Length (Metric) Cuff 5 cm