Learn More
MCR Safety Regular Weight Natural Cotton/Polyester Gloves with PVC Dots (1 Side)
Catalog No. 1904831C Shop All MCR Safety Products
Change view
Click to view available options
Large
Catalog No.
19-048-31C Large
1 options
This item is not returnable. View return policy
Catalog No. 19-048-31C Supplier MCR Safety Supplier No. 9650LM
This item is not returnable. View return policy
PVC dots on one side of the glove for increased grip, and abrasion resistance. Ideal for automotive, landscaping, and construction applications.
  • 7 Gauge Cotton/Polyester Blend
  • PVC Dots provide abrasion resistance and grip
Specifications
Construction String Knit
Coating Material PVC Dots (1 Side)
Color Natural
Hand Specific
Material Polycotton
Product Type String Knit Gloves
Size Large
Glove Cuff Color Natural
Glove Cuff Style Knit
Hem Color White
Packaging Type Polybag
Palm Type PVC Dots
Yarn Gauge 7
Length (English) 9.75 in.
Length (English) Cuff 2.75 in.
Length (Metric) 24.8 cm
Length (Metric) Cuff 7 cm