Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Regular Weight String Knit White Polyester Gloves

Catalog No. 19813576 Shop All MCR Safety Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-576 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-813-576 Supplier MCR Safety Supplier No. 9616LM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

7 gauge, white gloves are made with FDA accepted materials. Suitable for general hand protection in light duty applications.

  • 7 Gauge Regular Weight White
  • 100% Polyester Material
  • Made with FDA accepted materials
  • Reversible pattern

Specifications

Construction String Knit
Description Regular Weight
Color White
Hand Ambidextrous
Material Polyester
Product Type String Knit Gloves
Size Large
Style Reversible
Glove Cuff Color White
Glove Cuff Style Knit
Hem Color White
Packaging Type Polybag
Yarn Gauge 7
Length (English) 9.75 in.
Length (English) Cuff 2.75 in.
Length (Metric) 24.8 cm
Length (Metric) Cuff 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.