Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Regular Weight String Knit Cotton/Polyester Gloves with PVC Dots (2 Sides)

Catalog No. 19813580 Shop All MCR Safety Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-813-580 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-813-580 Supplier MCR Safety Supplier No. 9662LM
This item is not returnable. View return policy

Gloves feature a regular weight, 7 gauge gray cotton/polyester shell. PVC dots on both sides of the glove increase grip and abrasion resistance.

  • 7 gauge cotton and polyester work gloves
  • Black PVC dots on both sides
  • PVC dots provide more abrasion resistance and grip
  • 70% cotton/30% polyester blend
  • Comfortable, soft, and breathable
  • Color coded hem to identify sizes

Specifications

Construction String Knit
Coating Material PVC Dots (2 Sides)
Color Gray
Hand Ambidextrous
Material Polycotton
Product Type PVC-coated Gloves
Size Large
Glove Cuff Style Knit
Palm Type PVC Dots
Series 9662
Yarn Gauge 7
Length (English) 9.75 in.
Length (English) Cuff 2.75 in.
Length (Metric) 24.8 cm
Length (Metric) Cuff 7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.