Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Vinyl Coated General Purpose Work Gloves

Catalog No. 19054380 Shop All MCR Safety Products
Change view
Click to view available options
Small
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-054380 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-054380 Supplier MCR Safety Supplier No. 9880S
This item is not returnable. View return policy

Vinyl lamination offers abrasion resistance and an enhances wet/dry grip capabilities. A soft knit interlock cotton liner increases user comfort, while the 2 in. band top provides additional wrist protection.

  • Soft interlock knit lining stretches for a snug, comfortable fit
  • Gloves provide a high level of dexterity and tactile sensitivity
  • Vinyl material enhances abrasion resistance and wet/dry grip capabilities
  • 2 in. band top extends coverage up the wrist

Specifications

Coating Material Vinyl
Color Blue
Exterior Finish No Texture
Grade Industrial
Hand Specific
Product Type Dip-coated Gloves
Size Small
For Use With (Application) Automotive Assembly, and Warehousing
Glove Cuff Style Band Top
Liner Material Cotton
Series 9880
Thumb Style Wing
Length (English) 9.5 in.
Length (English) Cuff 2 in.
Length (Metric) 24 cm
Length (Metric) Cuff 5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.