Learn More
Description
- Dielectric, no metal parts
- Single lens design with integral side shields for greater orbital seal
- Spatula temples
- Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Compliance
ANSI Z87+ and CSA Z94.3
Specifications
Specifications
| Certifications/Compliance | ANSI Z87+, AS/NZS 1337.1,CE EN166,CAN/CSA Z94.3 |
| Description | 90% VLT |
| Lens Tint | Clear |
| Frame Color | Clear |
| No. per Pack | 144/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.