Learn More
Description
- Dielectric, no metal parts
- Single lens design with integral side shields for greater orbital seal
- Spatula temples
- Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Specifications
Specifications
| Certifications/Compliance | ANSI Z87+, CAN/CSA Z94.3 |
| Description | 90% VLT |
| Lens Tint | Clear |
| Frame Color | Clear |
| No. per Pack | 12/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.