Learn More
Description
Includes
Components of Kit
- Anti-phospho-MEK1 (Ser218/222)/MEK2 (Ser222/226)
- Raf-1 (truncated), active
- MEK1, unactive
- Magnesium/ATP Cocktail
- Assay Dilution Buffer I (ADBI)
Specifications
Specifications
| Detection Method | Chemiluminescence |
| For Use With (Application) | Kinase Assay |
| Includes | Anti-phospho-MEK1 (Ser218/222)/MEK2 (Ser222/226), Raf-1 (truncated), active, MEK1, unactive, Magnesium/ATP Cocktail, Assay Dilution Buffer I (ADBI) |
| Quantity | 1 Kit (50 Assays) |
| Type | Kinase Assay Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.