Learn More
Invitrogen™ MDC Monoclonal Antibody (D1), Invitrogen™
Description
Specifications
Specifications
| Antigen | MDC |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | CCL22 |
| Gene Accession No. | O00626 |
| Gene Alias | A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.