Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA527757
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA527757 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA527757 Supplier Invitrogen™ Supplier No. PA527757
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa nucleus. Predicted reactivity: Rhesus Monkey (96%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MDC1 participates in the early response to DNA damage. It is thought to be a member of the BRCT (BRCA1 C-terminus) super family of nuclear proteins. It contains an N-terminus forkhead-associated (FHA) motif, two C-terminus BRCT motifs and 13 internal repetitions of a 41 amino acid sequence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MDC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene MDC1
Gene Accession No. Q14676
Gene Alias 6820401C03; AA413496; DAAP-285E11.7; DKFZp781A0122; homologue to Drosophila photoreceptor protein calphotin; KIAA0170; MDC1; mediation of DNA damage checkpoint 1; mediator of DNA damage checkpoint 1; mediator of DNA damage checkpoint protein 1; mediator of DNA-damage checkpoint 1; MGC166888; mKIAA0170; NFBD1; nuclear factor with BRCT domains 1
Gene Symbols MDC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MDC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9656
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.