Learn More
Enzo Life Sciences Mdivi-1 (50mg). CAS: 338967-87-6
SDP
Supplier:  Enzo Life Sciences BMLCM1270050
Encompass_Preferred
Mdivi-1 is a selective inhibitor of mitochondrial division in yeast and mammalian cells which acts via inhibiting the mitochondrial division dynamin. In cells, Mdivi-1 inhibits apoptosis by inhibiting mitochondrial outer membrane permeabilization. Mdivi-1 causes the rapid (<5 min) reversible and dose-dependent formation of net-like mitochondria in wild-type cells with an IC50= ~10µM. In yeast, time-lapse fluorescence microscopy revealed no detectable mitochondrial division after treatment with Mdivi-1. It inhibits Dnm1 GTPase activity in a dose dependent manner with an estimated IC50 of 1-10µM but is not a general inhibitor of GTPases. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0434
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.