Learn More
Eurofins Discoverx MDL 29,951, 5 mg. (92-1264)
Supplier:  Eurofins Discoverx 921264
Encompass
MDL 29,951 is a synthetic small molecule selective agonist of the G protein-coupled receptor 17 (GPR17) that is active in intact cells. MDL 29,951 is selective over endogenous purinergic receptors. May be used in GPR17 studies and has a role in the maturation of oligodendrocytes. Provided as 5mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9783
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.