Learn More
Apexbio Technology LLC MDL-29951 130798-51-5 10mg
SDP
Supplier:  Apexbio Technology LLC B103810
Encompass_Preferred
MDL-29951 (CAS 130798-51-5) is a small molecule inhibitor targeting the glycine modulatory site of the N-methyl-D-aspartate (NMDA) receptor By antagonizing glycine-induced receptor activation MDL-29951 modulates NMDA receptor-mediated neurotransmission In vitro assays measuring [3H]glycine binding determined a Ki value of 0 14 mM indicating its affinity for the glycine site MDL-29951 is utilized in biomedical research to investigate the functional role of glycine co-agonism in NMDA receptor signaling and to explore the pharmacological modulation of glutamatergic pathways in neurological models
Catalog No. 50-000-94134
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.