Learn More
Santa Cruz Biotechnology MDM2 (SMP14)
Supplier:  Santa Cruz Biotechnology SC965
Encompass
MDM2 (SMP14), Source: Is a mouse monoclonal IgG1 antibody specific for the epitope corresponding to amino acids 154-167 of MDM2 of human origin, Molecular Weight of MDM2: 90 kDa, Storage: 4degreeC, do not freeze.
Catalog No. NC9539849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.