Learn More
Abcam MDR Assay Kit, Fluorometric, Quantitative, 1 x 96 Tests
Description
- Detection method: Fluorescent
- Sample types: Suspension cells and adherent cells
- Assay time: 1 hr
- Results type: Quantitative
- Measured target: P Glycoprotein
- Recommended instrument: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 96 Tests |
| Run Time | 1 hr. |
| Target Molecule | P-glycoprotein |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 96 Tests |
| For Use With (Application) | P-glycoprotein measurement |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.