Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam MDR Assay Kit, Fluorometric, Quantitative, 10x96 Tests
Description
- Assay Time: 1 hr
- Detection Method: Fluorescent
- Sample Types: Suspension cells and adherent cells
- Results Type: Quantitative
- Usage: Compatible with microplate readers
- Performance: Cited in > 6 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Run Time | 1 hr. |
| Target Molecule | P Glycoprotein |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 960 Tests |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 10 x 96 Tests |
| For Use With (Application) | P Glycoprotein measurement |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?