Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ MDR1 Vesicular Transport Assay Reagent Set

Catalog No. GM3030
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GM3030 100 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GM3030 Supplier Gibco™ Supplier No. GM3030
Season of Savings
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MDR1 Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except for the vesicles themselves.

The MDR1 Vesicular Transport Assay Reagent Set is a ready-to-use kit containing all reagents needed for vesicular transport assay except for the vesicles themselves. The reagent set can be used for vesicular transport assay using radioisotope-labeled or non-labeled compound. This kit has enough material to perform 100 vesicular transport assays.

Use The MDR1 Vesicular Transport Assay Reagent Set to:

  • Investigate the transporter interactions of your drug candidates
  • Assess potential for transporter-mediated drug-drug interactions
  • Obtain high quality results with a large signal to noise ratios

Concept Behind ABC Transporter Vesicles
ABC transporter vesicles are easy-to-use, efficient reagents for early assessment of a drug candidate's substrate and drug interaction potential. While ABC transporters typically mediate the export of substrates out of cells, transporters expressed on these inside-out vesicles import substrates into the vesicles. It is therefore possible to quantitatively evaluate transport activity for your compound by determining the amount incorporated into the vesicles.

Clear and Reliable Results
Prepared from Sf9 cells which have been engineered to over-express specific ABC transporters, these 'inside-out' vesicles provide high levels of transporter activity with low background, giving you a clear signal if your compound is a substrate or inhibitor of a specific efflux transporter.

Kit Compatibility
Our MDR1 Vesicular Transport Assay Reagent Set is designed for use with MDR1 Vesicles. The 100 vesicular transport assays that can be performed with this kit will require two MDR1 Vesicle products.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store Reagents A, B, C, and D at -20°C
Store Reagents E and F at room temperature
Quantity 100 Assays
Shipping Condition Dry Ice
Type Vesicular Transport Assay Reagent Set

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.