Learn More
Streck™ MDx-Chex™ for BCID2
SDP
Catalog No. 23285832 Shop All Streck Laboratories Products
Change view
Click to view available options
Includes:
2 BIOFIRE™ BCID2 Pouches
Catalog No. Includes
23-285-832 2 BIOFIRE™ BCID2 Pouches
1 options
This item is not returnable. View return policy
Catalog No. 23-285-832 Supplier Streck™ Supplier No. 250065
Add to Cart
Add to Cart
This item is not returnable. View return policy
MDx-Chex for BCID2 is a first-of-its-kind quality control designed to evaluate the entire analytical process of the BIOFIRE™ Blood Culture Identification 2 (BCID2) Panel for sepsis. With MDx-Chex for BCID2, you can track lot-to-lot performance of the BCID2 assay and reduce the occurrence of incorrect results due to instrument or assay failures.
  • Patient-like matrix of inactivated microorganisms, blood cells and culture media components.
  • Can be directly used on the BIOFIRE FilmArray™ Torch and 2.0 platforms.
  • Kit of two vials that provide complete coverage of all 43 BIOFIRE BCID2 Panel targets.
  • Challenges and verifies the entire analytical process of the BIOFIRE BCID2 Panel assay.
  • Reduces preanalytical variables such as pipetting errors and cross-contamination.
  • Easy to use - simply pipette like a patient sample.
Specifications
Type Quality Control
Includes 2 BIOFIRE™ BCID2 Pouches