Learn More
Streck MDx-Chex™ for Blood Culture - Gram Positive (BC-GP)
SDP
Catalog No. 23285982 Shop All Streck Laboratories Products
Change view
Click to view available options
Includes:
5 Positive Controls, 5 Negative Controls
Catalog No. Includes
23-285-982 5 Positive Controls, 5 Negative Controls
1 options
This item is not returnable. View return policy
Catalog No. 23-285-982 Supplier Streck Supplier No. 250079
Add to Cart
Add to Cart
This item is not returnable. View return policy
MDx-Chex for BC-GP and MDx-Chex for BC-GN are controls designed to evaluate the entire analytical process of the Luminex VERIGENE™ BC-GP and BC-GN Panels for sepsis. With MDx-Chex for BC-GP and MDx-Chex for BC-GN, you can track lot-to-lot performance of the BC-GP and BC-GN assays and reduce the occurrence of incorrect results due to instrument or assay failures.
  • Patient-like matrix of inactivated microorganisms, blood cells and culture media components.
  • Can be directly used on the Luminex VERIGENE system.
  • Complete coverage of all BC-GP or BC-GN Panel targets in a single vial.
  • Negative controls contain only blood cells and culture media components for a true patient-like negative control.
  • Challenges and verifies the entire analytical process of the Luminex VERIGENE assays.
  • Reduces preanalytical variables such as pipetting errors and cross-contamination.
Specifications
Type Quality Control
Includes 5 Positive Controls, 5 Negative Controls
Form Liquid