Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ME3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558408 Supplier Invitrogen™ Supplier No. PA558408
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVSLRIAIKV LDYAYKHNLA SYYPEPKDKE AFVRSLVYTP DYDSFTLDSY TWPKEAMNVQ TV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 76%.

ME3 proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis, metastasis, and atherosclerosis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ME3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ME3
Gene Accession No. Q16798
Gene Alias 1700020C08Rik; B230207H15Rik; HME; Macrophage elastase; malate dehydrogenase; malate dehydrogenase (oxaloacetate-decarboxylating) (NADP(+)); malic enzyme 3; malic enzyme 3, NADP(+)-dependent, mitochondrial; malic enzyme, NADP+-dependent, mitochondrial; ME; Me3; mitochondrial malic enzyme 3; mitochondrial NADP(+)-dependent malic enzyme 3; MME; MMP3; NADP-dependent malic enzyme, mitochondrial; NADP-ME; pyruvic-malic carboxylase
Gene Symbols ME3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ME3. Recombinant protein control fragment (Product #RP-97397).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10873
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.