Learn More
AdipoGen Me6BIO (5 mg)
SDP
Supplier:  AdipoGen AGMRC0021M005
Encompass_Preferred
Chemical. CAS: 667463-95-8. Formula: C17H12BrN3O2. MW: 370.2. N-methylated inactive control analog of 6-bromoindirubin-3'-oxime (6BIO) (Prod. No. MR-C0019). Displays minimal activity against CDK1/Cyclin B, GSK-3 alpha/beta and CDK5/p25. Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.
Catalog No. 50-205-4241
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.