Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mebendazole, Benzimidazole Antihelmintic, >99% Purity, Solid, 25 g

Catalog No. 502706767
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6767 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6767 Supplier Abcam Supplier No. AB14124625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzimidazole antihelmintic with antitumour activity in vitro and in vivo.

  • Molecular weight: 295.29 Da
  • Purity: >99%
  • Form: Solid
  • Class: Benzimidazole antihelmintic, tubulin-disrupting drug
  • Biological activity: Shows antitumour activity in vitro and in vivo; inhibits growth and induces apoptosis
  • Storage: Store at ambient temperature
  • Shipping: Ambient - can ship with ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Target Molecule Tubulin
Quantity 25 g
For Use With (Application) Anti-tumour research, Apoptosis, Tubulin-disruption
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.