Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mebendazole, Benzimidazole Antihelmintic, >99% Purity, Solid, 5 g

Catalog No. 502706766
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6766 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6766 Supplier Abcam Supplier No. AB1412465G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzimidazole antihelmintic with potent antitumour activity in vitro and in vivo.

  • Molecular Weight: 295.29 Da
  • Purity: >99%
  • Form: Solid
  • Activity: Benzimidazole antihelmintic and tubulin-disrupting drug with antitumour activity in vitro and in vivo; inhibits growth and induces apoptosis
  • Storage: Store at Ambient
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 100% Mebendazole

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Tubulin
Shipping Condition Ambient
Buffer 100% Mebendazole
Content And Storage Store at ambient temperature.
Form Solid
Formulation 100% Mebendazole
Quantity 5 g
For Use With (Application) Antitumour research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.