Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mecarbinate 98.0+%, TCI America™
SDP

Catalog No. M24071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24071G 1 g
M24075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24071G Supplier TCI America Supplier No. M24071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15574-49-9
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD00451373
InChI Key YTBNTDMBGXAOCG-UHFFFAOYSA-N
Synonym Ethyl 5-Hydroxy-1,2-dimethylindole-3-carboxylate, 5-Hydroxy-1,2-dimethylindole-3-carboxylic Acid Ethyl Ester
PubChem CID 616236
IUPAC Name ethyl 5-hydroxy-1,2-dimethylindole-3-carboxylate
SMILES CCOC(=O)C1=C(N(C2=C1C=C(C=C2)O)C)C

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 1 g
Formula Weight 233.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Mecarbinate
RTECSNumber NL6003600
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.