Learn More
Grainger Mech Expansion Plug Hex Nut 2.3 H 1 Pipe
Supplier:  Grainger 14M034
Encompass_Preferred
The Shaw Plugs 14M034 Mechanical Expansion Plug is a 3/8" diameter zinc alloy expansion anchor with a 1-1/4" embedment depth. It is designed for use in concrete and can support up to 75 lbs in tension and 225 lbs in shear. The expansion plug has a split end that expands outward when a screw is inserted to grip the sides of the hole tightly. It requires a 7/16" diameter hole and includes the screw for installation. This is a cost-effective concrete fastener for light-duty applications.
Catalog No. 50-275-1212
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.