Learn More
Grainger Mech Expansion Plug Snap Handle 2.61 H
Supplier:  Grainger 14M016
Encompass_Preferred
The Grainger 14M016 is a 14-inch diameter, 16-inch long, cast iron pulley with a 1-1/8 inch bore. It has a double groove for a V-belt and is rated for up to 15 horsepower at 1725 RPM. The pulley is made of durable cast iron with a black finish. It can be used for power transmission applications and with V-belt drives. The 1-1/8 inch bore allows it to fit on a variety of shaft sizes. This heavy-duty pulley provides reliable performance for industrial settings.
Catalog No. 50-275-2436
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.