Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ MechPro™ Thermal-Lined Waterproof Mechanics Gloves

Catalog No. 17200344 Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-200-344 Large
17-200-342 Small
17-200-345 X-Large
17-200-341 X-Small
17-200-343 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-200-344 Supplier Wells Lamont™ Supplier No. 7760L
Add to Cart
Add to Cart
This item is not returnable. View return policy

MechPro™ Thermal-Lined Waterproof Mechanics Gloves feature a spandex back for dexterity, and a goatskin leather palm for resistance to liquids. 3M™ Thinsulate™ lining provides additional comfort and thermal protection.

  • Premium goatskin leather palm
  • Gray spandex back increases dexterity
  • Neoprene knuckle strap absorbs shock
  • Waterproof membrane offers protection against liquids
  • 3M™ Thinsulate™ lining for cold environment applications
  • Feeltite™ hook and loop wrist closure

Specifications

Back Material Spandex
Color Black, Gray
Hand Specific
Material Goatskin Leather
Product Type Mechanic's Gloves
Size Large
Finger Style Full
For Use With (Application) Cold Weather, Refrigeration, Shipping, and Outdoor Construction
Glove Cuff Style Hook and Loop
Liner Material 3M™ Thinsulate™
Series 7760
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.