Learn More
Selleck Chemical LLC Meclizine 2HCl 50mg 1104-22-9 NSC 28728
SDP
Supplier:  Selleck Chemical LLC S198650MG
Encompass_Preferred
Meclizine (NSC 28728) is a histamine H1 receptor antagonist used to treat nausea and motion sickness, has anti-histamine, anti-muscarinic and anti-oxidative phosphorylation properties, also an agonist ligand for mCAR (constitutive androstane receptor) and an inverse agonist for hCAR. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1842
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.