Learn More
Description
Specifications
Specifications
| Concentration | 0.25mg/mL |
| Formulation | In 20mM Tris-HCl buffer (pH 8.0) containing 0.2M NaCl, 5mM DTT, 20% glycerol |
| Gene ID (Entrez) | 51102 |
| Molecular Weight (g/mol) | 36.9kDa |
| Purification Method | Protein |
| Quantity | 0.25mg |
| Source | Human |
| Immunogen | MGSSHHHHHH SSGLVPRGSH MPAKVVELKN LELAAVRGSD VRVKMLAAPI NPSDINMIQG NYGLLPELPA VGGNEGVAQV VAVGSNVTGL KPGDWVIPAN AGLGTWRTEA VFSEEALIQV PSDIPLQSAA TLGVNPCTAY RMLMDFEQLQ PGDSVIQNAS NSGVGQAVIQ IAAALGLRTI NVVRDRPDIQ KLSDRLKSLG AEHVITEEEL RRPEMKNFFK DMPQPRLALN CVGGKSSTEL LRQLARGGTM VTYGGMAKQP VVASVSLLIF KDLKLRGFWL SQWKKDHSPD QFKELILTLC DLIRRGQLTA PACSQVPLQD YQSALEASMK PFISSKQILT M |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
