Learn More
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1M Urea, pH 7.4. |
| Gene ID (Entrez) | 84246 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 26 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20C. Avoid freeze-thaw cycles. |
| Gene Alias | L6, mediator complex subunit 10TRG-17, mediator of RNA polymerase II transcription subunit 10, mediator of RNA polymerase II transcription, subunit 10, mediator of RNA polymerase II transcription, subunit 10 homolog (NUT2, S.cerevisiae), MGC5309, NUT2TRG-20, Transformation-related gene 17 protein, Transformation-related gene 20 protein, TRG20 |
| Gene Symbol | MED10 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-81328It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.For current lot information, including availability, please contact our technical support team click nb-technical@bio-techne.com For further blocking peptide related information and a protocol, click here . |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
