Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED19 Monoclonal Antibody (OTI2E6)
GREENER_CHOICE

Catalog No. PIMA527299
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527299 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527299 Supplier Invitrogen™ Supplier No. MA527299
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED19 Monoclonal Antibody for Western Blot

MED19 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED19
Applications Western Blot
Classification Monoclonal
Clone OTI2E6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MED19
Gene Accession No. A0JLT2
Gene Alias 2410018M14Rik; 3110040A13Rik; DT2P1G7; LCMR1; Lung cancer metastasis-related protein 1; Med19; MED19AS; mediator complex subunit 19; Mediator of RNA polymerase II transcription subunit 19; mediator of RNA polymerase II transcription, subunit 19 homolog; putative mediator subunit MED19AS protein; RGD1311926
Gene Symbols MED19
Host Species Mouse
Immunogen Full length human recombinant protein of MED19 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219541, 311165
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.