Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590677
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590677 Supplier Invitrogen™ Supplier No. PA590677
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGVTCVSQMP VAEGKSVQQT VELLTRKLEM LGAEKQGTFC VDCETYHTAA STLGSQGQTG KLMYVMHNSE YPLSCFALFE NGPCLIADTN FDVLMVKLKG FFQSAKASKI ETRGTRYQYC DFLVKVGTVT MGPSARGISV EVEYGPCVVA SDCWSLLLEF LQSFLGSHTP GAPAVFGNRH DAVYGPADTM VQYMELFNKI RKQQQVPVAG IR; Positive Samples: DU145, Mouse brain; Cellular Location: Nucleus.

MED20 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II, and general initiation factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MED20
Gene Accession No. Q5XIE9, Q9H944, Q9R0X0
Gene Alias 1110011O05Rik; 2410115I17Rik; AU018348; hTRFP; Med20; Mediator complex subunit 20; mediator of RNA polymerase II transcription subunit 20; PRO0213; Trf (TATA binding protein-related factor)-proximal homolog; Trf (TATA binding protein-related factor)-proximal protein homolog; TRFP; TRF-proximal protein homolog; ubiquitin specific protease 49; Usp49
Gene Symbols MED20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human MED20 (NP_0042662).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316209, 56771, 9477
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.