Learn More
Invitrogen™ MED22 Polyclonal Antibody

Description
Antibody detects endogenous levels of total MED22.
Specifications
Specifications
| Antigen | MED22 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | MED22 |
| Gene Accession No. | A0JPN6, Q15528, Q62276 |
| Gene Alias | AW212655; AW558812; MED22; MED24; Mediator complex subunit 22; mediator of RNA polymerase II transcription subunit 22; MGC48682; RGD1564893; si:ch73-278c20.1; si:dkey-226m8.2; surf5; Surf-5; surfeit 5; surfeit 5 isoform b; surfeit gene 5; surfeit locus protein 5; wu:fj45h07; zgc:86721 |
| Gene Symbols | MED22 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.