Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED26 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536140
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536140 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536140 Supplier Invitrogen™ Supplier No. PA536140
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 65 kDa. Purity is >95% by SDS-PAGE.

This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED26
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene MED26
Gene Accession No. O95402, Q7TN02
Gene Alias 5730493L18Rik; Activator-recruited cofactor 70 kDa component; AI414941; ARC70; AW495270; cofactor required for Sp1 transcriptional activation subunit 7; cofactor required for Sp1 transcriptional activation, subunit 7 (70kD); cofactor required for Sp1 transcriptional activation, subunit 7, 70kDa; CRSP complex subunit 7; Crsp7; CRSP70; Med26; Mediator complex subunit 26; mediator of RNA polymerase II transcription subunit 26; Slc35e1; solute carrier family 35, member E1; transcriptional coactivator CRSP70
Gene Symbols MED26
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1-50 of human CRSP70.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306328, 70625, 9441
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.