Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MED27 Monoclonal Antibody (OTI2A4)
GREENER_CHOICE

Catalog No. PIMA527129
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527129 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527129 Supplier Invitrogen™ Supplier No. MA527129
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED27 Monoclonal Antibody for Western Blot

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED27
Applications Western Blot
Classification Monoclonal
Clone OTI2A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MED27
Gene Accession No. Q6P2C8
Gene Alias 1500015J03Rik; 2310042P07Rik; AA682045; cofactor required for Sp1 transcriptional activation subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8; cofactor required for Sp1 transcriptional activation, subunit 8, 34kDa; CRAP34; CRSP complex subunit 8; CRSP34; CRSP8; D2Ertd434e; LOC525389 protein; MED27; MED3; mediator complex subunit 27; Mediator of RNA polymerase II transcription subunit 27; MGC11274; p37 TRAP/SMCC/PC2 subunit; RGD1564993; transcriptional coactivator CRSP34; transcriptional co-activator CRSP8 homolog; TRAP37
Gene Symbols MED27
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 24-247 of MED27 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9442
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.